Learn More
Grainger TRIPLETT Inspection Camera, 1920 x 1080
Supplier:  Grainger 792RJ7
Encompass_Preferred
The Triplett 1920 x 1080 Inspection Camera is a handheld digital inspection camera with a 1/4" CMOS sensor and six high-intensity LEDs. It captures still images and records video with 1920 x 1080 pixel resolution. The 3.5" LCD screen allows for live viewing. The camera comes with a 16.4' cable and has an IP67 waterproof and dustproof rating. It can store over 2000 photos on its 8GB SD card and is powered by a rechargeable lithium-ion battery. The camera has digital zoom, date/time stamp functions, and comes with USB and HDMI cables, as well as software for viewing images and videos on a PC.
Catalog No. NC2818015