Learn More
Electron Microscopy Sciences Tripod Polisher Model 590, Kit for TEM
SDP
Catalog No. 50980861
Encompass
The Tripod Polisher is used to prepare accurately micro sizes of TEM and SEM samples. For TEM samples, the Tripod Polisher iis used to limit ion milling times to less than 15 minutes, and in some cases, has eliminated the need for ion milling. It can be used to prepare both plan-view and cross-sections from a variety of sample materials, such as ceramics, composites, metals, and geological specimens. It has limited ion milling requirements that reduce preferential thinning, radiation damage, and heating of the sample. No strong chemicals are used to prepare the specimen.
Catalog No. 50-980-861 Supplier Electron Microscopy Sciences Supplier No. 5011510
The Tripod Polisher is used to prepare accurately micro sizes of TEM and SEM samples. For TEM samples, the Tripod Polisher iis used to limit ion milling times to less than 15 minutes, and in some cases, has eliminated the need for ion milling. It can be used to prepare both plan-view and cross-sections from a variety of sample materials, such as ceramics, composites, metals, and geological specimens. It has limited ion milling requirements that reduce preferential thinning, radiation damage, and heating of the sample. No strong chemicals are used to prepare the specimen.