Learn More
Chemglass Life Sciences Tripod Stand for 5 L Jacketed Bioreactor Vessel
SDP
Catalog No. 502733302
Encompass
Tripod stand designed to support 5 L jacketed bioreactor vessels. The top two-piece ring is anodized aluminum, while the lower ring and legs are electropolished stainless steel, offering stable, corrosion-resistant support.
  • Supports 5 L jacketed bioreactor vessels.
  • Top two-piece ring is anodized aluminum.
  • Lower ring and legs are electropolished stainless steel.
  • Includes silicone o-ring and six knurled nuts for headplate attachment.
  • Provides stable support for bioprocess applications.
Catalog No. 50-273-3302 Supplier Chemglass Life Sciences Supplier No. CLS1386STD5
Tripod stand designed to support 5 L jacketed bioreactor vessels. The top two-piece ring is anodized aluminum, while the lower ring and legs are electropolished stainless steel, offering stable, corrosion-resistant support.
  • Supports 5 L jacketed bioreactor vessels.
  • Top two-piece ring is anodized aluminum.
  • Lower ring and legs are electropolished stainless steel.
  • Includes silicone o-ring and six knurled nuts for headplate attachment.
  • Provides stable support for bioprocess applications.