Learn More
Chemglass Life Sciences Chemglass Tripod Stand for 5 L Unjacketed Bioreactor Vessels
SDP
Catalog No. NC2892884
Encompass
The Chemglass Tripod Stand is designed to support 5 L unjacketed bioreactor vessels, providing a stable, corrosion-resistant base for laboratory bioprocessing and cell culture workflows.
  • Supports 5 L unjacketed bioreactor vessels.
  • Anodized aluminum top two-piece ring for secure vessel seating.
  • Electropolished stainless steel lower ring and legs for corrosion resistance.
  • Includes a silicone o-ring and six knurled nuts for headplate attachment.
  • Compact footprint for benchtop use; dimensions not specified by manufacturer.
Catalog No. NC2892884 Supplier Chemglass Life Sciences Supplier No. CLS1382STD5
The Chemglass Tripod Stand is designed to support 5 L unjacketed bioreactor vessels, providing a stable, corrosion-resistant base for laboratory bioprocessing and cell culture workflows.
  • Supports 5 L unjacketed bioreactor vessels.
  • Anodized aluminum top two-piece ring for secure vessel seating.
  • Electropolished stainless steel lower ring and legs for corrosion resistance.
  • Includes a silicone o-ring and six knurled nuts for headplate attachment.
  • Compact footprint for benchtop use; dimensions not specified by manufacturer.