Learn More
Chemglass Life Sciences Tripod Stand for 7L Unjacketed Bio Reactor Vessels
SDP
Catalog No. 501294471
Encompass
Tripod Stand for 7L Unjacketed Bio Reactor Vessels. Stand to hold unjacketed bio reaction vessels. Top, two-piece ring is made of anodized aluminum while the lower ring and legs are electropolished stainless steel. Stands come complete with silicone o-ring and six, knurled nuts to attach the headplate to the stand.Like-for-like replacement parts for Applikon Biotechnology and Broadley-James brands of bioreactors.Applikon is a registered trademark of Applikon Biotechnology, Inc. Broadley-James and Broadley-James Corporation are registered trademarks of Broadley-James Corporation.
Catalog No. 50-129-4471 Supplier Chemglass Life Sciences Supplier No. CLS1383STD7
Tripod Stand for 7L Unjacketed Bio Reactor Vessels. Stand to hold unjacketed bio reaction vessels. Top, two-piece ring is made of anodized aluminum while the lower ring and legs are electropolished stainless steel. Stands come complete with silicone o-ring and six, knurled nuts to attach the headplate to the stand.Like-for-like replacement parts for Applikon Biotechnology and Broadley-James brands of bioreactors.Applikon is a registered trademark of Applikon Biotechnology, Inc. Broadley-James and Broadley-James Corporation are registered trademarks of Broadley-James Corporation.