Learn More
AdipoGen Tripolin A (1 mg)
SDP
Supplier:  AdipoGen AGCR13520M001
Encompass_Preferred
Chemical. CAS: 1148118-92-6. Formula: C15H11NO3. MW: 253.3. Specific non-ATP competitive Aurora A kinase inhibitor. Does not significantly inhibit Aurora B kinase in mammalian cells. Reduces localization of pAurora A on spindle microtubules (MTs), affects centrosome integrity, spindle formation and length and MT dynamics in interphase.
Catalog No. 50-205-3903