Learn More
Ted Pella Inc 400ml Tri-Stir Beakers, 20/pk
SDP
Catalog No. NC9000311
Encompass
Convenient measuring and stirring of EM chemicals, pours without dripping, a disposable alternative to your laboratory glassware. Polypropylene Tri-Stir Disposable Beakers are graduated, stackable, disposable, sterilizable and have three shallow dripless pouring spouts. The Tri-Stir Disposable Plastic Beaker is useful for EM/LM specimen preparation. The Tri-Stir beakers are also called Tri-Pour and Tri-Corner beakers. Polypropylene Tri-Stir Disposable Beakers have a service temperature up to 100°C (212°F).
Catalog No. NC9000311 Supplier Ted Pella Inc Supplier No. 1290820
Convenient measuring and stirring of EM chemicals, pours without dripping, a disposable alternative to your laboratory glassware. Polypropylene Tri-Stir Disposable Beakers are graduated, stackable, disposable, sterilizable and have three shallow dripless pouring spouts. The Tri-Stir Disposable Plastic Beaker is useful for EM/LM specimen preparation. The Tri-Stir beakers are also called Tri-Pour and Tri-Corner beakers. Polypropylene Tri-Stir Disposable Beakers have a service temperature up to 100°C (212°F).