Learn More
Andwin Scientific TRIPOUR BEAKER CAP 100ML 500PK
SDP
Catalog No. 50727615
Encompass_Preferred
The wax impregnated Tripout Beaker Cap is to be used with the 100ml Tricorn Beaker. It features lift tabs that may be written on for identification. It is sold in a pack of 500.
Catalog No. 50-727-615 Supplier Andwin Scientific Supplier No. 60971
The wax impregnated Tripout Beaker Cap is to be used with the 100ml Tricorn Beaker. It features lift tabs that may be written on for identification. It is sold in a pack of 500.