Learn More
Electron Microscopy Sciences Tri-Pour Beaker Cap for 50 ml beaker
SDP
Catalog No. 501898983
Encompass_Preferred
The caps are available for all Tri-Pour beakers. The caps are wax impregnated, have lift-tabs, and may be written on for sample identification.
Use: 50 ml
Catalog No. 50-189-8983 Supplier Electron Microscopy Sciences Supplier No. 60969
The caps are available for all Tri-Pour beakers. The caps are wax impregnated, have lift-tabs, and may be written on for sample identification.
Use: 50 ml