Learn More
Enterprise Technology Solutions Tripp Lite Adapter Cable, USB-C to VGA, Gen 1, USB 3.1, M/M, 6'L, Black
SDP
Catalog No. 5000739689
Encompass_Preferred
USB 3.1, generation 1 USB-C to VGA 4K adapter cable (M/M) helps you transmit video from your tablet, laptop, Chromebook, MacBook, smartphone or PC to a VGA-enabled TV, monitor or projector. Plug-and-play technology makes installation quick and simple - with no software or drivers to install. Fumble-free, reversible USB-C plug connects in either direction for a quick connection every time. Foil and braid shielding protects against EMI/RFI line noise that can distort or destroy data. Integral strain relief helps cable and connectors move freely without stress or cracking for lasting use. 6' cable supports HD video resolutions up to 1920 x 1200 (including 1080p) at 60 Hz and 36-bit Deep Color (12 bits per channel). Cable is perfect for giving video presentations in conference rooms and classrooms, editing multiple documents on a larger screen, or streaming video for digital signage in crystal-clear 1080p.
Catalog No. 50-007-39689 Supplier Enterprise Technology Solutions Supplier No. TRPU444006V
USB 3.1, generation 1 USB-C to VGA 4K adapter cable (M/M) helps you transmit video from your tablet, laptop, Chromebook, MacBook, smartphone or PC to a VGA-enabled TV, monitor or projector. Plug-and-play technology makes installation quick and simple - with no software or drivers to install. Fumble-free, reversible USB-C plug connects in either direction for a quick connection every time. Foil and braid shielding protects against EMI/RFI line noise that can distort or destroy data. Integral strain relief helps cable and connectors move freely without stress or cracking for lasting use. 6' cable supports HD video resolutions up to 1920 x 1200 (including 1080p) at 60 Hz and 36-bit Deep Color (12 bits per channel). Cable is perfect for giving video presentations in conference rooms and classrooms, editing multiple documents on a larger screen, or streaming video for digital signage in crystal-clear 1080p.