Learn More
Enterprise Technology Solutions Tripp Lite Cat5e Cat5 Straight Through Modular Shielded In-Line Coupler RJ45 F/F
SDP
Catalog No. 502111054
Encompass_Preferred
Tripp Lite's Shielded Cat5e standalone RJ45 straight inline coupler provides a female-to-female, feed-through connection. It allows you to extend the length of a Cat5e patch cable by adding another Cat5e patch cable. Shielded housing protects against EMI/RFI. UL certified.
Catalog No. 50-211-1054 Supplier Enterprise Technology Solutions Supplier No. N032001
Tripp Lite's Shielded Cat5e standalone RJ45 straight inline coupler provides a female-to-female, feed-through connection. It allows you to extend the length of a Cat5e patch cable by adding another Cat5e patch cable. Shielded housing protects against EMI/RFI. UL certified.