Learn More
Enterprise Technology Solutions Tripp Lite Converter Adapter, DisplayPort 1.2 to VGA/DVI/HDMI, Black
SDP
Catalog No. 5000740436
Encompass_Preferred
DisplayPort 1.2 to VGA/DVI/HDMI converter adapter connects your laptop or tablet's DisplayPort to a VGA, DVI or HDMI display. The all-in-one unit contains separate female DVI, HD15 and HDMI ports. Plug-and-play technology makes installation quick and simple - with no software or drivers to install. Converter adapter supports video resolutions up to 1920 x 1080 (DVI), and 1920 x 1200, including 1080p (HD15) and 3840 x 2160 (HDMI). It also supports 36-bit Deep Color (12 bits per channel). The passive adapter requires the connected computer to have a DP++ (DisplayPort++) port. Converter adapter is compatible with laptops, tablets and other devices equipped with a female DisplayPort. With its small size and built-in cable, this unit is perfect for stowing in your laptop bag and carrying to school, the office or the latest trade show.
Catalog No. 50-007-40436 Supplier Enterprise Technology Solutions Supplier No. TRPP13606NHDV4K
DisplayPort 1.2 to VGA/DVI/HDMI converter adapter connects your laptop or tablet's DisplayPort to a VGA, DVI or HDMI display. The all-in-one unit contains separate female DVI, HD15 and HDMI ports. Plug-and-play technology makes installation quick and simple - with no software or drivers to install. Converter adapter supports video resolutions up to 1920 x 1080 (DVI), and 1920 x 1200, including 1080p (HD15) and 3840 x 2160 (HDMI). It also supports 36-bit Deep Color (12 bits per channel). The passive adapter requires the connected computer to have a DP++ (DisplayPort++) port. Converter adapter is compatible with laptops, tablets and other devices equipped with a female DisplayPort. With its small size and built-in cable, this unit is perfect for stowing in your laptop bag and carrying to school, the office or the latest trade show.