Learn More
Enterprise Technology Solutions Tripp Lite Converter Adapter, mDP 1.2 to VGA/DVI/HDMI, All-in-One, BK
SDP
Catalog No. 5000740014
Encompass_Preferred
Mini DisplayPort 1.2 to VGA/DVI/HDMI converter adapter connects your laptop or tablet's Mini DisplayPort or Thunderbolt 1/2 port to a VGA, DVI or HDMI display. Plug-and-play technology makes installation quick and simple - with no software or drivers to install. The all-in-one unit contains separate female DVI, HD15 and HDMI ports. Converter adapter supports video resolutions up to 1920 x 1080 (DVI), and 1920 x 1200, including 1080p (HD15), and 3840 x 2160 (HDMI). It also supports 36-bit Deep Color (12 bits per channel). The passive adapter requires the connected computer to have a DP++ (DisplayPort++) port. With its small size and built-in cable, this unit is perfect for stowing in your laptop bag and carrying to school, the office or the latest trade show.
Catalog No. 50-007-40014 Supplier Enterprise Technology Solutions Supplier No. TRPP13706NHDV4K
Mini DisplayPort 1.2 to VGA/DVI/HDMI converter adapter connects your laptop or tablet's Mini DisplayPort or Thunderbolt 1/2 port to a VGA, DVI or HDMI display. Plug-and-play technology makes installation quick and simple - with no software or drivers to install. The all-in-one unit contains separate female DVI, HD15 and HDMI ports. Converter adapter supports video resolutions up to 1920 x 1080 (DVI), and 1920 x 1200, including 1080p (HD15), and 3840 x 2160 (HDMI). It also supports 36-bit Deep Color (12 bits per channel). The passive adapter requires the connected computer to have a DP++ (DisplayPort++) port. With its small size and built-in cable, this unit is perfect for stowing in your laptop bag and carrying to school, the office or the latest trade show.