Learn More
Enterprise Technology Solutions Tripp Lite Economy Surge Protector, 6 Outlet, 790 Joules, 6' Cord, WE
SDP
Catalog No. 5000739349
Encompass_Preferred
Surge protector provides economical AC surge suppression for workstations, desktop PCs, telecom systems and other sensitive electronics. Built-in AC surge suppression shields connected equipment from damage and performance problems due to transient surges. TLP604 offers six total outlets, 15 amp resettable circuit breaker and a 6' cord. AC suppression rated at 790 joules shields equipment from surges and line noise. It includes full normal mode (H-N) and common mode (N-G/H-G) line surge suppression. Attractive gray housing with keyhole mounting tabs and one designated transformer plug outlet offers convenient protection of desktop computer equipment and accessories.
Catalog No. 50-007-39349 Supplier Enterprise Technology Solutions Supplier No. TRPTLP606
Surge protector provides economical AC surge suppression for workstations, desktop PCs, telecom systems and other sensitive electronics. Built-in AC surge suppression shields connected equipment from damage and performance problems due to transient surges. TLP604 offers six total outlets, 15 amp resettable circuit breaker and a 6' cord. AC suppression rated at 790 joules shields equipment from surges and line noise. It includes full normal mode (H-N) and common mode (N-G/H-G) line surge suppression. Attractive gray housing with keyhole mounting tabs and one designated transformer plug outlet offers convenient protection of desktop computer equipment and accessories.