Learn More
Enterprise Technology Solutions Tripp Lite HDMI Cable, w/ Ethernet, Ultra HD 4Kx2K, 16'L, Black
SDP
Catalog No. 5000740225
Encompass_Preferred
HDMI cable with Ethernet supports Ethernet Data Connectivity, Audio Return Channel, 3D support and video resolutions up to 4096 x 2160. High-speed cable has been tested to perform at speeds of 340 Mhz or up to 18 Gbps and can successfully handle 1080p signals including those at increased color depths and increased refresh rates from the source. Gold-plated connectors provide optimal signal transfer for higher quality. HDMI cable is backwards-compatible with previous versions of HDMI and will work in all HDMI cabling scenarios. 16' cable is perfect for connecting an iPad2 to an HDTV (video mirroring) to watch movies, play games, surf the web and display presentations.
Catalog No. 50-007-40225 Supplier Enterprise Technology Solutions Supplier No. TRPP569016
HDMI cable with Ethernet supports Ethernet Data Connectivity, Audio Return Channel, 3D support and video resolutions up to 4096 x 2160. High-speed cable has been tested to perform at speeds of 340 Mhz or up to 18 Gbps and can successfully handle 1080p signals including those at increased color depths and increased refresh rates from the source. Gold-plated connectors provide optimal signal transfer for higher quality. HDMI cable is backwards-compatible with previous versions of HDMI and will work in all HDMI cabling scenarios. 16' cable is perfect for connecting an iPad2 to an HDTV (video mirroring) to watch movies, play games, surf the web and display presentations.