Learn More
Enterprise Technology Solutions Tripp Lite Line-Interactive 2U Rack/Tower UPS, 8-Out, 1500 VA, BK
SDP
Supplier:  Enterprise Technology Solutions TRPSMC15002URM
Encompass_Preferred
Line-interactive, uninterruptible power supply (UPS) with enhanced LCD interface offers network-grade power protection for your critical server, network and telecommunications equipment. UPS with built-in, auto-voltage regulation (AVR) actively corrects brownouts and overvoltages back to usable levels while maintaining a full battery charge in case of a power failure. Interactive LCD interface reports UPS operating mode, detailed UPS and site-power data, plus enables a variety of UPS setup and configuration options. Super-fast switchover from line to battery power occurs within milliseconds to maintain operation of connected equipment without interruption. 94 percent line-mode efficiency offers reduced heat emissions and operating costs. USB and RS-232 ports work with PowerAlert software to communicate detailed UPS status and site power conditions. Communication ports enable detailed monitoring of equipment load levels, self-test data and utility power conditions.
Catalog No. 50-007-40369