Learn More
Enterprise Technology Solutions Tripp Lite Patch Cable, Category 6, 24 Wire Gauge, 1'L, Gray
SDP
Catalog No. 5000740052
Encompass_Preferred
This category 6, molded patch cable lets you stay ahead of the game by using a high-speed cable to distribute data, voice and video. Cable delivers stable performance up to 550MHz. High-quality copper wire and a staggered-pin plug design keep near-end cross-talk levels to a minimum. Snagless design protects the locking tabs on the RJ45 connectors from being damaged or snapped off during installation. 1' cable is designed for use in category 6, category 5e and category 5 applications.
Catalog No. 50-007-40052 Supplier Enterprise Technology Solutions Supplier No. TRPN201001GY
This category 6, molded patch cable lets you stay ahead of the game by using a high-speed cable to distribute data, voice and video. Cable delivers stable performance up to 550MHz. High-quality copper wire and a staggered-pin plug design keep near-end cross-talk levels to a minimum. Snagless design protects the locking tabs on the RJ45 connectors from being damaged or snapped off during installation. 1' cable is designed for use in category 6, category 5e and category 5 applications.