Learn More
Enterprise Technology Solutions Tripp Lite Surge Protector, 10-Outlet, 5-7/10"Wx11-1/10"Lx1-3/10"H, BK
SDP
Catalog No. 5000740442
Encompass_Preferred
10-outlet surge protector features a surge protection rating of 2880 joules to defend your sensitive electronic components against strong surges and spikes. Surge protector includes full normal mode (H-N) and common mode (N-G/H-G) line surge suppression as well as technology that filters out EMI/RFI line noise to prevent interference from disrupting your electronics and inflicting damage or data loss. NEMA 5-15R outlets are widely spaced to accept bulky transformers without blocking adjacent outlets. Safety covers keep dust and debris away from unused outlets. Integrated 15A circuit breaker protects all of the outlets and shuts down your connected equipment in case of overload. On/off switch provides one-touch control over your connected equipment at once. Diagnostic LED bars indicate power and protection status at a glance. The 6' AC power cord with right-angle NEMA 5-15P plug allows furniture and equipment to be placed flush against the wall.
Catalog No. 50-007-40442 Supplier Enterprise Technology Solutions Supplier No. TRPTLP1006B
10-outlet surge protector features a surge protection rating of 2880 joules to defend your sensitive electronic components against strong surges and spikes. Surge protector includes full normal mode (H-N) and common mode (N-G/H-G) line surge suppression as well as technology that filters out EMI/RFI line noise to prevent interference from disrupting your electronics and inflicting damage or data loss. NEMA 5-15R outlets are widely spaced to accept bulky transformers without blocking adjacent outlets. Safety covers keep dust and debris away from unused outlets. Integrated 15A circuit breaker protects all of the outlets and shuts down your connected equipment in case of overload. On/off switch provides one-touch control over your connected equipment at once. Diagnostic LED bars indicate power and protection status at a glance. The 6' AC power cord with right-angle NEMA 5-15P plug allows furniture and equipment to be placed flush against the wall.