Learn More
Enterprise Technology Solutions Tripp Lite Surge Protector,790Joules,LEDLight,6Outlet,6ftCord,White
SDP
Catalog No. 5000740441
Encompass_Preferred
Protect It! Six-Outlet Surge Protector offers economical AC surge suppression to protect workstations, desktop PCs, telecom systems and other sensitive electronics. Built-in AC surge suppression shields connected equipment from damage and performance problems due to transient surges. Automatic shutoff permanently cuts power to outlets if protection circuit is incapacitated, preventing equipment damage and indicating where replacement is required. LED light warns of suppressor damage. AC suppression, rated at 790 joules, shields equipment from surges and line noise. Attractive housing with keyhole mounting tabs offer convenient protection of desktop equipment and accessories.
Catalog No. 50-007-40441 Supplier Enterprise Technology Solutions Supplier No. TRPTLP606TAA
Protect It! Six-Outlet Surge Protector offers economical AC surge suppression to protect workstations, desktop PCs, telecom systems and other sensitive electronics. Built-in AC surge suppression shields connected equipment from damage and performance problems due to transient surges. Automatic shutoff permanently cuts power to outlets if protection circuit is incapacitated, preventing equipment damage and indicating where replacement is required. LED light warns of suppressor damage. AC suppression, rated at 790 joules, shields equipment from surges and line noise. Attractive housing with keyhole mounting tabs offer convenient protection of desktop equipment and accessories.