Learn More
Enterprise Technology Solutions Tripp Lite UPS,Cloud-Connected,350VA,210-Watt,3 Outlets,BK
SDP
Catalog No. 5000739850
Encompass_Preferred
350VA battery backup for PCs and retail point-of-sale application is a standby, Cloud-connected UPS that provides reliable battery backup and AC power protection against blackouts, brownouts, overvoltages, surges and line noise. 210-watt, 120-volt UPS protects home/office computing, networking and point-of-sale equipment in retail, kiosks, foodservice and light industrial environments. It's easy to set up in minutes. Just plug it into a properly grounded wall outlet. Press the power button and connect equipment to the UPS outlets. Eaton's cloud-based, remote, monitoring dashboard offers convenient setup, control and management of a single UPS. Receive alerts, push configurations, silence alarms or control outlet banks using a mobile app or web browser. Unit's NFC area lets you transfer custom network settings instantly from your mobile device. UPS features three NEMA 5-15R outlets.
Catalog No. 50-007-39850 Supplier Enterprise Technology Solutions Supplier No. TRPBC350RNC
350VA battery backup for PCs and retail point-of-sale application is a standby, Cloud-connected UPS that provides reliable battery backup and AC power protection against blackouts, brownouts, overvoltages, surges and line noise. 210-watt, 120-volt UPS protects home/office computing, networking and point-of-sale equipment in retail, kiosks, foodservice and light industrial environments. It's easy to set up in minutes. Just plug it into a properly grounded wall outlet. Press the power button and connect equipment to the UPS outlets. Eaton's cloud-based, remote, monitoring dashboard offers convenient setup, control and management of a single UPS. Receive alerts, push configurations, silence alarms or control outlet banks using a mobile app or web browser. Unit's NFC area lets you transfer custom network settings instantly from your mobile device. UPS features three NEMA 5-15R outlets.