Learn More
CDW Eaton Tripp Lite Series UPS Smart LCD 1500VA 900W 120V Line-Interactive UPS - 8 Outlets USB DB9 2U Rack/Tower Battery
Catalog No. NC3595917
Encompass_Preferred
Eaton Tripp Lite Series UPS Smart LCD 1500VA 900W 120V Line-Interactive UPS - 8 Outlets USB DB9 2U Rack/Tower Battery
Catalog No. NC3595917 Supplier CDW Supplier No. 892647
Eaton Tripp Lite Series UPS Smart LCD 1500VA 900W 120V Line-Interactive UPS - 8 Outlets USB DB9 2U Rack/Tower Battery