Learn More
TrippNT TRIPPNT LabFlow Tray, Polypropylene, 6 in. Depth, Translucent
SDP
Catalog No. NC3997674 Shop All TrippNT Products
Encompass
This LabFlow Tray provides durable, chemical-resistant storage with translucent visibility and secure containment. Designed for LabFlow carts and racks, it keeps supplies organized and easily accessible. Its deep 6 in. design is ideal for bulky storage and offers high visibility of contents.
  • Deep 6 in. tray for bulky or high-volume supply storage
  • Translucent polypropylene walls allow instant visibility of contents
  • Lightweight, chemical-resistant polypropylene construction
  • Provides containment, portability, and easy segregation of supplies
  • Made in the U.S.A.
  • Modular design provides a flexible foundation for storage systems, compatible with all carts and racks, simplifying organization and speeding up supply access
  • Corrosion resistant
  • BPA free and chemical free
  • NSF listed
  • -40°F to 160°F temperature range
Catalog No. NC3997674 Supplier TrippNT Supplier No. 101170
This LabFlow Tray provides durable, chemical-resistant storage with translucent visibility and secure containment. Designed for LabFlow carts and racks, it keeps supplies organized and easily accessible. Its deep 6 in. design is ideal for bulky storage and offers high visibility of contents.
  • Deep 6 in. tray for bulky or high-volume supply storage
  • Translucent polypropylene walls allow instant visibility of contents
  • Lightweight, chemical-resistant polypropylene construction
  • Provides containment, portability, and easy segregation of supplies
  • Made in the U.S.A.
  • Modular design provides a flexible foundation for storage systems, compatible with all carts and racks, simplifying organization and speeding up supply access
  • Corrosion resistant
  • BPA free and chemical free
  • NSF listed
  • -40°F to 160°F temperature range