Learn More
Selleck Chemical LLC Triprolidine Hydrochloride S5447-1g
SDP
Catalog No. 503110219
Encompass_Preferred
Triprolidine Hydrochloride is the hydrochloride salt form of Triprolidine which is the first generation histamine H1 antagonist used in allergic rhinitis
Catalog No. 50-311-0219 Supplier Selleck Chemical LLC Supplier No. S54471G
Triprolidine Hydrochloride is the hydrochloride salt form of Triprolidine which is the first generation histamine H1 antagonist used in allergic rhinitis