Learn More
Medchemexpress LLC Triprolidine (hydrochloride) | 550-70-9 | 25MG
SDP
Catalog No. 5000413839
Encompass_Preferred
Triprolidine hydrochloride is an orally active histamine H1 antagonist. It has the function of spinal cord motor and sensory block and can be used for the research of allergic rhinitis.
  • Orally active histamine H1 antagonist.
  • Exhibits spinal cord motor and sensory block function.
  • Can antagonize histamine H1 and decrease the expression of CD45 in maturing human dendritic cells.
  • Produces a dose-dependent effect of spinal motor and sensory block in rats when administered intrathecally.
Catalog No. 50-004-13839 Supplier Medchemexpress LLC Supplier No. 25MGHYB1808A
Triprolidine hydrochloride is an orally active histamine H1 antagonist. It has the function of spinal cord motor and sensory block and can be used for the research of allergic rhinitis.
  • Orally active histamine H1 antagonist.
  • Exhibits spinal cord motor and sensory block function.
  • Can antagonize histamine H1 and decrease the expression of CD45 in maturing human dendritic cells.
  • Produces a dose-dependent effect of spinal motor and sensory block in rats when administered intrathecally.