Learn More
Medchemexpress LLC Triprolidine (hydrochloride) | 550-70-9 | MFCD00039044 | 99.8% | 314.85 g/mol | C19H23ClN2 | 10 MG
SDP
Catalog No. 5000298124
Encompass_Preferred
Triprolidine hydrochloride is a first-generation antihistamine and an orally active histamine H1 receptor antagonist supplied for research use. It is provided as a white to off-white solid with high reported purity and is available in small-milligram pack sizes for laboratory studies.
  • First-generation H1 receptor antagonist for research applications.
  • High purity suitable for analytical and in vitro assays.
  • Available in small mg quantities for screening and dose-response studies.
  • White to off-white solid, easy to weigh and dissolve in common solvents.
  • Store sealed away from moisture; recommended storage: 4°C for solid, in solvent: -80°C (6 months) or -20°C (1 month).
  • For research use only; not for human or clinical use.
Catalog No. 50-002-98124 Supplier Medchemexpress LLC Supplier No. HYB1808A10MG
Triprolidine hydrochloride is a first-generation antihistamine and an orally active histamine H1 receptor antagonist supplied for research use. It is provided as a white to off-white solid with high reported purity and is available in small-milligram pack sizes for laboratory studies.
  • First-generation H1 receptor antagonist for research applications.
  • High purity suitable for analytical and in vitro assays.
  • Available in small mg quantities for screening and dose-response studies.
  • White to off-white solid, easy to weigh and dissolve in common solvents.
  • Store sealed away from moisture; recommended storage: 4°C for solid, in solvent: -80°C (6 months) or -20°C (1 month).
  • For research use only; not for human or clinical use.