Learn More
Medchemexpress LLC Triprolidine hydrochloride | 550-70-9 | MFCD00039044 | 99.8% | 5 MG
SDP
Catalog No. 5000412630
Encompass_Preferred
Triprolidine hydrochloride is an orally active histamine H1 antagonist. It functions as a spinal cord motor and sensory block and is used for research related to allergic rhinitis.
  • Orally active histamine H1 antagonist
  • Functions as a spinal cord motor and sensory block
  • Used in research for allergic rhinitis
Catalog No. 50-004-12630 Supplier Medchemexpress LLC Supplier No. HYB1808A5MG
Triprolidine hydrochloride is an orally active histamine H1 antagonist. It functions as a spinal cord motor and sensory block and is used for research related to allergic rhinitis.
  • Orally active histamine H1 antagonist
  • Functions as a spinal cord motor and sensory block
  • Used in research for allergic rhinitis