Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Triprolidine Hydrochloride, British Pharmacopoeia (BP) Reference Standard, MilliporeSigma™ Supelco™

Catalog No. 111022891 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-102-2891 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-102-2891 Supplier MilliporeSigma Supelco Supplier No. BP347
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets, has been developed and issued under the Authority of the issuing Pharmacopoeia.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Pharmacopeia & Metrological Institutes Standards
CAS 6138-79-0
Flash Point Not Applicable
Molecular Formula C19H22N2 · HCl
Linear Formula C19H22N2 · HCl
Quantity 100 mg
MDL Number MFCD00039044
Synonym (E)-2-[3-(1-Pyrrolidinyl)-1-p-tolylpropenyl]pyridine hydrochloride; trans-2-[3-(1-Pyrrolidinyl)-1-p-tolylpropenyl]pyridine hydrochloride
Formula Weight 314.85
Grade BP
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.