Learn More
TARGETMOL CHEMICALS INC Triprolidine hydrochloride mon
SDP
Catalog No. 502878184
Encompass_Preferred
Also available in 1 mL, 10 mg, 25 mg, 50 mg, 500 mg and bulk. Please contact Fisher for quotes. Triprolidine hydrochloride monohydrate is a histamine H1 antagonist used in allergic rhinitis, asthma. Purity 98%
Catalog No. 50-287-8184 Supplier TARGETMOL CHEMICALS INC Supplier No. T1030100MG
Also available in 1 mL, 10 mg, 25 mg, 50 mg, 500 mg and bulk. Please contact Fisher for quotes. Triprolidine hydrochloride monohydrate is a histamine H1 antagonist used in allergic rhinitis, asthma. Purity 98%