Learn More
Medchemexpress LLC Triprolidine (hydrochloride monohydrate) | 6138-79-0 | 99.9% | 1 ML
SDP
Catalog No. 5000412504
Encompass_Preferred
Triprolidine hydrochloride monohydrate is a first-generation antihistamine that acts as an oral active histamine H1 antagonist. It can be utilized in research concerning allergic rhinitis and has demonstrated spinal motor and sensory block in rats.
  • First-generation antihistamine.
  • Oral active histamine H1 antagonist.
  • Applicable for allergic rhinitis research.
  • Exhibits spinal motor and sensory block in rats.
Catalog No. 50-004-12504 Supplier Medchemexpress LLC Supplier No. HYB130110MM/1ML
Triprolidine hydrochloride monohydrate is a first-generation antihistamine that acts as an oral active histamine H1 antagonist. It can be utilized in research concerning allergic rhinitis and has demonstrated spinal motor and sensory block in rats.
  • First-generation antihistamine.
  • Oral active histamine H1 antagonist.
  • Applicable for allergic rhinitis research.
  • Exhibits spinal motor and sensory block in rats.