Learn More
Medchemexpress LLC Triprolidine hydrochloride monohydrate | 6138-79-0 | >99.9% | 25 MG
SDP
Catalog No. 5000413840
Encompass_Preferred
Triprolidine hydrochloride monohydrate is a first-generation antihistamine that acts as an oral active histamine H1 antagonist. It is typically used for research purposes as an inhibitor.
  • Acts as a histamine H1 antagonist.
  • Suitable for research applications.
  • High purity for reliable results.
  • Available in a 25 MG quantity.
Catalog No. 50-004-13840 Supplier Medchemexpress LLC Supplier No. 25MGHYB1301
Triprolidine hydrochloride monohydrate is a first-generation antihistamine that acts as an oral active histamine H1 antagonist. It is typically used for research purposes as an inhibitor.
  • Acts as a histamine H1 antagonist.
  • Suitable for research applications.
  • High purity for reliable results.
  • Available in a 25 MG quantity.