Learn More
Medchemexpress LLC Triprolidine hydrochloride monohydrate | 6138-79-0 | MFCD00150574 | 99.9% | 332.87 | C19H25ClN2O | 10 MG
SDP
Catalog No. 5000295931
Encompass_Preferred
Triprolidine hydrochloride monohydrate is a first-generation histamine H1 receptor antagonist provided as a research reagent for in vitro and related laboratory studies. It is supplied as a solid monohydrate for solution preparation and analytical work.
  • First-generation H1 histamine antagonist used for pharmacological research.
  • Provided as a solid monohydrate, white to off-white in appearance.
  • Molecular formula C19H25ClN2O and molecular weight 332.87 g/mol.
  • Available in small measured packs suitable for laboratory assays and preparative work.
  • Typical quality assurance includes batch COA with purity determined by LCMS.
Catalog No. 50-002-95931 Supplier Medchemexpress LLC Supplier No. HYB130110MG
Triprolidine hydrochloride monohydrate is a first-generation histamine H1 receptor antagonist provided as a research reagent for in vitro and related laboratory studies. It is supplied as a solid monohydrate for solution preparation and analytical work.
  • First-generation H1 histamine antagonist used for pharmacological research.
  • Provided as a solid monohydrate, white to off-white in appearance.
  • Molecular formula C19H25ClN2O and molecular weight 332.87 g/mol.
  • Available in small measured packs suitable for laboratory assays and preparative work.
  • Typical quality assurance includes batch COA with purity determined by LCMS.