Learn More
Medchemexpress LLC Tripropyl phosphate-d21 | 1219794-92-9 | 98.0% | 245.36 | 25 MG
SDP
Catalog No. 5000452658
Encompass_Preferred
Tripropyl phosphate-d21 is a deuterium labeled version of Tripropyl phosphate. It is intended for research use only and can be used as a tracer or an internal standard for quantitative analysis. Deuterium substitution has shown potential to influence the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-004-52658 Supplier Medchemexpress LLC Supplier No. 25MGHYW012351S
Tripropyl phosphate-d21 is a deuterium labeled version of Tripropyl phosphate. It is intended for research use only and can be used as a tracer or an internal standard for quantitative analysis. Deuterium substitution has shown potential to influence the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.