Learn More
Medchemexpress LLC Tripropylammonium hexafluorophosphate | 92416-28-9 | MFCD00035574 | 289.24 g/mol | C9H22F6NP | 250mg
SDP
Catalog No. 5000270217
Encompass_Preferred
Tripropylammonium hexafluorophosphate is a potent cell-permeable free radical scavenger suitable for the detection of superoxide radicals and peroxynitrite in vitro
Catalog No. 50-002-70217 Supplier Medchemexpress LLC Supplier No. HYW790758250MG
Tripropylammonium hexafluorophosphate is a potent cell-permeable free radical scavenger suitable for the detection of superoxide radicals and peroxynitrite in vitro