Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tripropylene glycol, 99%, pure, mixture of isomers

Catalog No. AC458480050
Change view
Click to view available options
Quantity:
1 L
5 L
25 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC458480050 5 L
AC458480010 1 L
AC458480250 25 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC458480050 Supplier Thermo Scientific Chemicals Supplier No. 458480050
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 24800-44-0 | C9H20O4 | 192.25 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 24800-44-0
Molecular Formula C9H20O4
Molecular Weight (g/mol) 192.25

Specifications

Density 1.0200g/mL
Boiling Point 270.0°C
Flash Point 145°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (sum of isomers) (GC)
Quantity 5 L
Specific Gravity 1.02
Solubility Information Solubility in water: soluble.
Viscosity 77.3 mm²/s (20°C)
Formula Weight 192.25
Percent Purity 99%
Grade Pure
Chemical Name or Material Tripropylene glycol
Show More Show Less
EINECSNumber 246-466-

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.