Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tripropylphosphine, 98%, Thermo Scientific™

Catalog No. AC383410050
Change view
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC383410050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC383410050 Supplier Thermo Scientific Chemicals Supplier No. 383410050

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2234-97-1
Molecular Formula C9H21P
Molecular Weight (g/mol) 160.241
MDL Number MFCD00015217
InChI Key KCTAHLRCZMOTKM-UHFFFAOYSA-N
Synonym tripropylphosphine, tri-propylphosphine, tri-n-propylphosphine, phosphine, tripropyl, tri-n-propylphosphine,min, tri n-propyl phosphine, acmc-1cipf, ppr n 3, tripropyl-phosphane
PubChem CID 75226
IUPAC Name tripropylphosphane
SMILES CCCP(CCC)CCC

Specifications

Color Colorless
Density 0.8010g/mL
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Refractive Index 1.4570 to 1.4590 (20°C, 589nm)
Linear Formula [CH3(CH2)2]3P
Quantity 5 g
Formula Weight 160.24
Percent Purity ≥97.5%
Physical Form Liquid
Chemical Name or Material Tripropylphosphine
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes serious eye damage.
Harmful if swallowed.
Causes severe skin burns and eye damage.
Harmful in contact with skin.
Catches fire spontaneously if exposed to air.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Do not allow contact with air.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.