Learn More
Medchemexpress LLC Celastrol | 34157-83-0 | MFCD03424073 | >98.0% | C29H36O4 | 10MG
SDP
Catalog No. 501872800
Encompass_Preferred
Celastrol is a natural triterpenoid small molecule used in research to inhibit proteasome activity and to induce autophagy and apoptosis. It is applied in cellular and biochemical studies investigating proteostasis, inflammatory signaling, and metabolic regulation.
  • High purity (>98%).
  • Solid powder suitable for storage and formulation.
  • Potent proteasome inhibitor and autophagy inducer.
  • Applicable to cellular and biochemical assays.
  • Available in multiple pack sizes and solution formats for experimental flexibility.
  • COA and SDS available to support quality and safety documentation.
Catalog No. 50-187-2800 Supplier Medchemexpress LLC Supplier No. HY1306710MG
Celastrol is a natural triterpenoid small molecule used in research to inhibit proteasome activity and to induce autophagy and apoptosis. It is applied in cellular and biochemical studies investigating proteostasis, inflammatory signaling, and metabolic regulation.
  • High purity (>98%).
  • Solid powder suitable for storage and formulation.
  • Potent proteasome inhibitor and autophagy inducer.
  • Applicable to cellular and biochemical assays.
  • Available in multiple pack sizes and solution formats for experimental flexibility.
  • COA and SDS available to support quality and safety documentation.