Learn More
Cayman Chemical Wilforgine, Purity: ≥98%, CAS: 37239-47-7, 10 mg
SDP
Catalog No. 502910517
Encompass_Preferred
A sesquiterpene alkaloid with paralytic activity; induces paralysis in third instar M. separata larvae (NC50 = 0.0086 µmol/larvae)
Catalog No. 50-291-0517 Supplier Cayman Chemical Supplier No. 3502910 MG
A sesquiterpene alkaloid with paralytic activity; induces paralysis in third instar M. separata larvae (NC50 = 0.0086 µmol/larvae)