Learn More
Medchemexpress LLC HY-32735 2mg , Triptolide CAS:38748-32-2 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY327352MG
Encompass_Preferred
HY-32735 2mg Triptolide CAS: 38748-32-2 Triptolide is a diterpenoid triepoxide extracted from the root of Tripterygium wilfordii with immunosuppressive, anti-inflammatory, antiproliferative and antitumour effects. Triptolide is a NF-κB activation inhibitor[4][5][6]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-9272