Learn More
American Radiolabeled Chemicals Inc TRIPTOLIDE 3HG 50UC
SDP
Catalog No. 50353007
Encompass_Preferred
Triptolide-3H(G) 50uCi in ethanol, 1 mCi/ml, 10-20 Ci/mmol, M.W. 360.4, Shipped in Dry Ice, Exclusive
Catalog No. 50-353-007 Supplier American Radiolabeled Chemicals Inc Supplier No. ART 200850UC
Add to Cart
Add to Cart
Request a Quote
Triptolide-3H(G) 50uCi in ethanol, 1 mCi/ml, 10-20 Ci/mmol, M.W. 360.4, Shipped in Dry Ice, Exclusive