Learn More
Cayman Chemical TrIptolIde-d3 1mg
SDP
Catalog No. 502910542
Encompass_Preferred
An internal standard for the quantification of triptolide by GC- or LC-MS
Catalog No. 50-291-0542 Supplier Cayman Chemical Supplier No. 348951 MG
An internal standard for the quantification of triptolide by GC- or LC-MS