Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Triptolide-d3 | 38748-32-2 | MFCD00210565 | 91.9% | 363.42 g·mol⁻¹ | C20H21D3O6 | 1 MG
SDP

Catalog No. 5000284823
Encompass_Preferred

Triptolide-d3 (d3) is the deuterium-labeled isotopologue of triptolide supplied as a stable isotope standard for analytical and research applications. It is intended for use as an internal standard or tracer in mass spectrometry and isotopic studies, and is provided in a 1 mg package.

  • Deuterium-labeled (d3) isotopologue for internal standard use in mass spectrometry
  • Purity 91.9% (>90%)
  • Molecular formula C20H21D3O6
  • Molecular weight 363.42 g·mol⁻¹
  • Package size 1 mg
  • Store at -20°C; protect from light; handle under inert atmosphere where indicated

Catalog No. 50-002-84823 Supplier Medchemexpress LLC Supplier No. HYW6543771MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Triptolide-d3 (d3) is the deuterium-labeled isotopologue of triptolide supplied as a stable isotope standard for analytical and research applications. It is intended for use as an internal standard or tracer in mass spectrometry and isotopic studies, and is provided in a 1 mg package.

  • Deuterium-labeled (d3) isotopologue for internal standard use in mass spectrometry
  • Purity 91.9% (>90%)
  • Molecular formula C20H21D3O6
  • Molecular weight 363.42 g·mol⁻¹
  • Package size 1 mg
  • Store at -20°C; protect from light; handle under inert atmosphere where indicated

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.