Learn More
Medchemexpress LLC Triptolide palmitate | 2126920-51-0 | >98.0% | 598.81 g/mol | C36H54O7 | 10 MG
SDP
Catalog No. 5000288007
Encompass_Preferred
Triptolide palmitate is the palmitate ester derivative of triptolide provided as a research reagent. It is a lipophilic compound supplied as an oil and characterized for in vitro cytotoxicity and preliminary pharmacokinetic properties; it is intended for laboratory research use only and not for human use or clinical applications.
  • Derivative of triptolide, designed to increase lipophilicity and suitability for formulation.
  • Demonstrated cytotoxicity to MCF-7 and A549 cancer cell lines (IC50 ≈ 7.5 μM and 6.4 μM).
  • Supplied as an oil with high DMSO solubility (100 mg/mL) for easy stock preparation.
  • Includes storage guidance: store at 4°C protected from light; in solution, keep at -80°C (6 months) or -20°C (1 month).
  • Provided for research use only; not intended for patient or clinical use.
Catalog No. 50-002-88007 Supplier Medchemexpress LLC Supplier No. HY15809010MG
Triptolide palmitate is the palmitate ester derivative of triptolide provided as a research reagent. It is a lipophilic compound supplied as an oil and characterized for in vitro cytotoxicity and preliminary pharmacokinetic properties; it is intended for laboratory research use only and not for human use or clinical applications.
  • Derivative of triptolide, designed to increase lipophilicity and suitability for formulation.
  • Demonstrated cytotoxicity to MCF-7 and A549 cancer cell lines (IC50 ≈ 7.5 μM and 6.4 μM).
  • Supplied as an oil with high DMSO solubility (100 mg/mL) for easy stock preparation.
  • Includes storage guidance: store at 4°C protected from light; in solution, keep at -80°C (6 months) or -20°C (1 month).
  • Provided for research use only; not intended for patient or clinical use.