Learn More
Selleck Chemical LLC Triptolide (PG490) 5mg 38748-32-2 NSC 163062
SDP
Supplier:  Selleck Chemical LLC S36045MG
Encompass_Preferred
Triptolide (PG490, NSC 163062) is a diterpene triepoxide, immunosuppresive agent extracted from the Chinese herb Tripterygium wilfordii. It functions as a NF-?B inhibitor with dual actions by disruption of p65/CBP interaction and by reduction of p65 protein. Triptolide (PG490) abrogates the transactivation function of heat shock transcription factor 1 (HSF1). Triptolide inhibits MDM2 and induces apoptosis through a p53-independent pathway. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. NC1344592