Learn More
Cayman Chemical TrIptophenolIde 10mg
SDP
Catalog No. 502948520
Encompass_Preferred
A diterpenoid with antiandrogenic activity; an antagonist of wild-type and mutant ARs (IC50s = 260, 388, 480, and 437 nM for wild-type, ART877A, ARF876L, and ARW741C/T877A, respectively, in transactivation assays); inhibits the growth of, and reduces AR levels in, LNCaP prostate cancer cells at 5 µM
Catalog No. 50-294-8520 Supplier Cayman Chemical Supplier No. 3439010 MG
A diterpenoid with antiandrogenic activity; an antagonist of wild-type and mutant ARs (IC50s = 260, 388, 480, and 437 nM for wild-type, ART877A, ARF876L, and ARW741C/T877A, respectively, in transactivation assays); inhibits the growth of, and reduces AR levels in, LNCaP prostate cancer cells at 5 µM