Learn More
Bioworld TRIPTORELIN (RECOMBINANT)
Supplier:  Bioworld 220607351
Encompass_Preferred
Triptorelin Acetate Source: Description : Triptorelin C64H82N18O13, Pyr-His-Trp-Ser-Tyr-D-Trp-Leu-Arg-Pro-Gly-NH2 is a synthetic analogue of gonadorelin (GnRH). As a result of the substitution of the 6th amino acid residue in the ...
Catalog No. 50-253-833