Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Triptycene 98.0+%, TCI America™
SDP

Catalog No. T28071G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T28071G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T28071G Supplier TCI America Supplier No. T28071G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 477-75-8
Molecular Formula C20H14
Molecular Weight (g/mol) 254.33
MDL Number MFCD00003813
InChI Key NGDCLPXRKSWRPY-UHFFFAOYSA-N
Synonym triptycene, tryptycene, unii-cl32869mep, 9,10 1',2'-benzenoanthracene, 9,10-dihydro, 9,10-o-benzeno-9,10-dihydroanthracene, 9,10-dihydro-9,10-1,2 benzenoanthracene, tribenzobicyclo 2.2.2 octatriene, 9,10-dihydro-9,10-o-benzenoanthracene
PubChem CID 92764
IUPAC Name pentacyclo[6.6.6.0²,⁷.0⁹,¹⁴.0¹⁵,²⁰]icosa-2,4,6,9,11,13,15,17,19-nonaene
SMILES C1=CC=C2C3C4=CC=CC=C4C(C2=C1)C1=CC=CC=C31

Specifications

Melting Point 255°C
Color White-Yellow
Quantity 1 g
Formula Weight 254.33
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Triptycene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.