Learn More
Sigma Aldrich Fine Chemicals Biosciences TriPureTM Isolation Reagen
Catalog No. 502905301
Encompass
Monophasic SOLUTIONon of phenol and guanidine thiocyanate for RNA DNA and protein isolation.
Catalog No. 50-290-5301 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 11667165001
Monophasic SOLUTIONon of phenol and guanidine thiocyanate for RNA DNA and protein isolation.