Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Triruthenium Dodecacarbonyl

Catalog No. 2450111G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
5 g
50 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
24-501-11G 1 g
24-501-150G 50 g
24-501-15G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Warning
Catalog No. 24-501-11G Supplier Sigma Aldrich Supplier No. 2450111G
May include imposed supplier surcharges.
Add to Cart
Add to Cart
Warning

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Atomic number of base material: 44 RutheniumCatalyst used in a [3+2+1] carbonylative cycloaddition of silylacteylenes and +a,+â-unsaturated ketones providing good yields of +a-pyrones.A Practical Procedure for Reduction of Primary, Secondary and Tertiary Amides to Amines

Specifications

CAS 15243-33-1
Molecular Formula C12O12Ru3
Linear Formula Ru3(CO)12
MDL Number MFCD00011209
Quantity 1 g
Synonym Ruthenium carbonyl; tri-Ruthenium dodecacarbonyl
Molecular Weight (g/mol) 639.33 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.