Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals TRIS, 0.5M buffer soln., pH 6.8

Catalog No. AAJ63735AP
Change view
Click to view available options
Quantity:
500 mL
1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ63735AP 500 mL
AAJ63735K2 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ63735AP Supplier Thermo Scientific Chemicals Supplier No. J63735AP
Add to Cart

CAS: 77-86-1 | C4H11NO3 | 121.136 g/mol

TRIS is used as a standard biological buffer in less critical applications where economy is an important consideration, component of extraction buffer e.g. phenol extraction of DNA or RNA; and as a component of separating and stacking gels in the characterization of in vitro translation products by SDS-PAGE.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
TRIS is used as a standard biological buffer in less critical applications where economy is an important consideration, component of extraction buffer e.g. phenol extraction of DNA or RNA; and as a component of separating and stacking gels in the characterization of in vitro translation products by SDS-PAGE.

Solubility
Difficult to mix.

Notes
Store in cool, dry conditions in well sealed containers. Keep container tightly closed.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 77-86-1
Molecular Formula C4H11NO3
Molecular Weight (g/mol) 121.136
MDL Number MFCD00004679
InChI Key LENZDBCJOHFCAS-UHFFFAOYSA-N
PubChem CID 6503
ChEBI CHEBI:9754
IUPAC Name 2-amino-2-(hydroxymethyl)propane-1,3-diol
SMILES C(C(CO)(CO)N)O

Specifications

Concentration 0.5 M
pH 6.8
Physical Form Liquid
Quantity 500 mL
Solubility Information Difficult to mix.
Chemical Name or Material TRIS buffer soln.
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.