Learn More
Medchemexpress LLC Tris(1,10-phenanthroline) ruthenium(II) chloride | 23570-43-6 | MFCD00061374 | ≥95.0% | 712.59 g/mol | C36H24Cl2N6Ru | 250 MG
SDP
Supplier:  Medchemexpress LLC HYW145116250MG
Encompass_Preferred
Tris(1,10-phenanthroline) ruthenium(II) chloride is a photosensitive ruthenium complex used as a photocatalyst for light-driven organic transformations and photoelectrochemical studies. It is supplied for research use and supports visible-light-mediated reactions.
  • Photosensitive photocatalyst for visible-light mediated reactions.
  • Suitable for organic synthesis and photoelectrochemical applications.
  • High purity, typically ≥95.0% as listed by suppliers.
  • Brown solid with good stability when protected from light.
  • Intended for research use only; follow safety data sheet guidance.
Catalog No. 50-002-83775